Merck
  • Cortical Neural Stem Cell Lineage Progression Is Regulated by Extrinsic Signaling Molecule Sonic Hedgehog.
Cortical Neural Stem Cell Lineage Progression Is Regulated by Extrinsic Signaling Molecule Sonic Hedgehog.
Cell reports (2020-04-03)
Yue Zhang, Guoping Liu, Teng Guo, Xiaoyi G Liang, Heng Du, Lin Yang, Aparna Bhaduri, Xiaosu Li, Zhejun Xu, Zhuangzhi Zhang, Zhenmeiyu Li, Miao He, Jeremiah Tsyporin, Arnold R Kriegstein, John L Rubenstein, Zhengang Yang, Bin Chen
ZUSAMMENFASSUNG
Neural stem cells (NSCs) in the prenatal neocortex progressively generate different subtypes of glutamatergic projection neurons. Following that, NSCs have a major switch in their progenitor properties and produce γ-aminobutyric acid (GABAergic) interneurons for the olfactory bulb (OB), cortical oligodendrocytes, and astrocytes. Herein, we provide evidence for the molecular mechanism that underlies this switch in the state of neocortical NSCs. We show that, at around E16.5, mouse neocortical NSCs start to generate GSX2-expressing (GSX2+) intermediate progenitor cells (IPCs). In vivo lineage-tracing study revealed that GSX2+ IPC population gives rise not only to OB interneurons but also to cortical oligodendrocytes and astrocytes, suggesting that they are a tri-potential population. We demonstrated that Sonic hedgehog signaling is both necessary and sufficient for the generation of GSX2+ IPCs by reducing GLI3R protein levels. Using single-cell RNA sequencing, we identify the transcriptional profile of GSX2+ IPCs and the process of the lineage switch of cortical NSCs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Olig-2-Antikörper, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Olig2-Antikörper, Klon 211F1.1, clone 211F1.1, from mouse
Sigma-Aldrich
Anti-Gsh2-Antikörper, serum, from rabbit
Sigma-Aldrich
ANTI-GSX2 (C-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution