Merck
Rapid in vitro production of single-stranded DNA.
Nucleic acids research (2019-11-13)
Dionis Minev, Richard Guerra, Jocelyn Y Kishi, Cory Smith, Elisha Krieg, Khaled Said, Amanda Hornick, Hiroshi M Sasaki, Gabriel Filsinger, Brian J Beliveau, Peng Yin, George M Church, William M Shih
ZUSAMMENFASSUNG
There is increasing demand for single-stranded DNA (ssDNA) of lengths >200 nucleotides (nt) in synthetic biology, biological imaging and bionanotechnology. Existing methods to produce high-purity long ssDNA face limitations in scalability, complexity of protocol steps and/or yield. We present a rapid, high-yielding and user-friendly method for in vitro production of high-purity ssDNA with lengths up to at least seven kilobases. Polymerase chain reaction (PCR) with a forward primer bearing a methanol-responsive polymer generates a tagged amplicon that enables selective precipitation of the modified strand under denaturing conditions. We demonstrate that ssDNA is recoverable in ∼40-50 min (time after PCR) with >70% yield with respect to the input PCR amplicon, or up to 70 pmol per 100 μl PCR reaction. We demonstrate that the recovered ssDNA can be used for CRISPR/Cas9 homology directed repair in human cells, DNA-origami folding and fluorescent in-situ hybridization.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammoniumpersulfat, Molecular Biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
Natriumacrylat, 97%
Sigma-Aldrich
N,N′-Dimethylethylendiamin, 98%