Merck
  • Artemisinin and dihydroartemisinin promote β-cell apoptosis induced by palmitate via enhancing ER stress.
Artemisinin and dihydroartemisinin promote β-cell apoptosis induced by palmitate via enhancing ER stress.
Apoptosis : an international journal on programmed cell death (2020-01-03)
Ke Chen, Hu Hua, Ziyang Zhu, Tong Wu, Zhanjun Jia, Qianqi Liu
ZUSAMMENFASSUNG
Artemisinin (ART) and dihydroartemisinin (DHA) are first-line antimalarial drugs and have been reported to have anti-obesity effects. Hyperlipidemia is associated with β-cell damage in obese subjects, which could contribute to the pathogenesis of type 2 diabetes. In addition to their anti-obesity effects, ART and DHA also have protective roles in some diseases. Thus, we investigated the effects of ART and DHA in palmitate-induced β-cell apoptosis and the underlying mechanism. In this study, the rat pancreatic β-cell line INS-1 and mouse pancreatic β-cell line MIN6 were cultured with palmitate (PA) (0.1 mM) to induce cell apoptosis in the presence or absence of ART or DHA. Cell apoptosis was investigated by using flow cytometry, and the expression of ER stress markers, including CHOP, GRP78 and PDI, was detected by Western blotting and/or qRT-PCR. The results showed that ART and DHA significantly increased the apoptosis of β-cells induced by PA and exacerbated the ER stress caused by PA. An inhibitor of ER stress, 4-phenylbutyric acid (4-PBA), significantly ameliorated cell apoptosis caused by ART and DHA in PA-treated β-cells, consistent with the inhibition of ER stress. Together, the findings from the current study suggested that ART and DHA may promote lipid disorder-associated β-cell injury via enhancing ER stress when they were used to treat obesity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin-Fragment 26-33 Amid, ≥97% (HPLC), powder