Merck
Creation of (R)-Amine Transaminase Activity within an α-Amino Acid Transaminase Scaffold.
ACS chemical biology (2020-01-29)
Moritz Voss, Chao Xiang, Jérémy Esque, Alberto Nobili, Marian J Menke, Isabelle André, Matthias Höhne, Uwe T Bornscheuer
ZUSAMMENFASSUNG
The enzymatic transamination of ketones into (R)-amines represents an important route for accessing a range of pharmaceuticals or building blocks. Although many publications have dealt with enzyme discovery, protein engineering, and the application of (R)-selective amine transaminases [(R)-ATA] in biocatalysis, little is known about the actual in vivo role and how these enzymes have evolved from the ubiquitous α-amino acid transaminases (α-AATs). Here, we show the successful introduction of an (R)-transaminase activity in an α-amino acid aminotransferase with one to six amino acid substitutions in the enzyme's active site. Bioinformatic analysis combined with computational redesign of the d-amino acid aminotransferase (DATA) led to the identification of a sextuple variant having a specific activity of 326 milliunits mg-1 in the conversion of (R)-phenylethylamine and pyruvate to acetophenone and d-alanine. This value is similar to those of natural (R)-ATAs, which typically are in the range of 250 milliunits mg-1. These results demonstrate that (R)-ATAs can evolve from α-AAT as shown here for the DATA scaffold.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
α-Methylbenzylamin, 99%
Sigma-Aldrich
(±)-2-(p-Methoxyphenoxy)propionic acid, ≥98%