Merck
  • Micheliolide ameliorates diabetic kidney disease by inhibiting Mtdh-mediated renal inflammation in type 2 diabetic db/db mice.
Micheliolide ameliorates diabetic kidney disease by inhibiting Mtdh-mediated renal inflammation in type 2 diabetic db/db mice.
Pharmacological research (2019-11-02)
Wenting Liu, Xiaowen Chen, Yuxian Wang, Yihua Chen, Sijia Chen, Wangqiu Gong, Ting Chen, Lingzhi Sun, Chenghao Zheng, Bohui Yin, Shuting Li, Congwei Luo, Qianyin Huang, Jing Xiao, Zhaozhong Xu, Fenfen Peng, Haibo Long
ZUSAMMENFASSUNG
Diabetic kidney disease (DKD) is the principal cause of end-stage renal disease worldwide and few treatments are available. Because immunomodulators are pivotal to DKD pathophysiology, anti-inflammatory agents may be useful for treating DKD. This study was conducted to investigate the effect of micheliolide (MCL), a novel guaianolide sesquiterpene lactone with well-known anti-inflammatory effects, on DKD. Treatment with dimethylaminomicheliolide (DMAMCL), the pro-drug of MCL currently under clinical trial in oncology, protected the kidneys against proteinuria, renal failure, histopathological injury, and inflammation in db/db mice. This effect was associated with metadherin (Mtdh) downregulation. We observed aberrant upregulation of Mtdh in the kidneys of db/db mice and high-glucose (HG)-induced mouse tubular epithelial cells (mTECs). Downregulation of Mtdh obviously inhibited nuclear factor-κB signaling activation and suppressed its downstream inflammatory cytokines, such as monocyte chemotactic peptide-1, interleukin-1β, tumor necrosis factor-α, and interleukin-6 in HG-induced mTECs, which was similar to the effect of MCL. Mtdh overexpression largely reversed the anti-inflammatory role of MCL. Moreover, MCL downregulated Mtdh by both inhibiting the transcription level and promoting ubiquitin-mediated degradation. These findings suggest that DMAMCL is a promising anti-inflammatory agent useful for preventing renal injury in DKD by inhibiting Mtdh-mediated renal inflammation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)