Merck
  • Cancer Cells Resist Mechanical Destruction in Circulation via RhoA/Actomyosin-Dependent Mechano-Adaptation.
Cancer Cells Resist Mechanical Destruction in Circulation via RhoA/Actomyosin-Dependent Mechano-Adaptation.
Cell reports (2020-03-19)
Devon L Moose, Benjamin L Krog, Tae-Hyung Kim, Lei Zhao, Sophia Williams-Perez, Gretchen Burke, Lillian Rhodes, Marion Vanneste, Patrick Breheny, Mohammed Milhem, Christopher S Stipp, Amy C Rowat, Michael D Henry
ZUSAMMENFASSUNG
During metastasis, cancer cells are exposed to potentially destructive hemodynamic forces including fluid shear stress (FSS) while en route to distant sites. However, prior work indicates that cancer cells are more resistant to brief pulses of high-level FSS in vitro relative to non-transformed epithelial cells. Herein, we identify a mechano-adaptive mechanism of FSS resistance in cancer cells. Our findings demonstrate that cancer cells activate RhoA in response to FSS, which protects them from FSS-induced plasma membrane damage. We show that cancer cells freshly isolated from mouse and human tumors are resistant to FSS, that formin and myosin II activity protects circulating tumor cells (CTCs) from destruction, and that short-term inhibition of myosin II delays metastasis in mouse models. Collectively, our data indicate that viable CTCs actively resist destruction by hemodynamic forces and are likely to be more mechanically robust than is commonly thought.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
SAFC
Insulin, human, rekombinant, for research or for further manufacturing use, dry powder
Sigma-Aldrich
Anti-β-Actin-Antikörper, Maus monoklonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Oleoyl-L-α-lysophosphatidische Säure Natriumsalz, ≥98%, solid
Sigma-Aldrich
Ionomycin aus Streptomyces conglobatus, ≥98% (HPLC)
Sigma-Aldrich
CK-666, ≥98% (HPLC), powder
Sigma-Aldrich
Arp2/3-Komplex-Inhibitor I, CK-666, Arp2/3 Complex Inhibitor I, CK-666, CAS 442633-00-3, is a cell-permeable selective inhibitor of actin assembly mediated by actin-related protein Arp2/3 complex (IC50 = 4 uM in human).
Sigma-Aldrich
ML-7, powder