Merck
  • Design and in vitro evaluation of electrospun shape memory polyurethanes for self-fitting tissue engineering grafts and drug delivery systems.
Design and in vitro evaluation of electrospun shape memory polyurethanes for self-fitting tissue engineering grafts and drug delivery systems.
Materials science & engineering. C, Materials for biological applications (2020-03-25)
Monika Bil, Ewa Kijeńska-Gawrońska, Eliza Głodkowska-Mrówka, Aneta Manda-Handzlik, Piotr Mrówka
ZUSAMMENFASSUNG
Integration of multiple features including shape memory, biodegradation, and sustained drug delivery in a single material offers the opportunity to significantly improve the abilities of implantable devices for cardiovascular system regeneration. Two types of shape memory polyurethanes (SMPUs): PU-PLGA and PU-PLLA/PEG differing in soft segments composition that comprising blends of various biodegradable polyols, i.e. D,l-lactide-co-glycolide diol (o-PLGA), poly(e-caprolactone) diols (o-PCL) with various molecular weights, poly-l-lactide diol (o-PLLA), polyethylene glycol (o-PEG) were synthesized and further utilized to electrospun nanofibrous - rapamycin (Rap) delivery system. Structure characterization by Fourier transform infrared spectroscopy (FTIR), differential scanning calorimetry (DCS) and hydrophilicity measurements were performed to gain more insights on the influence of the particular units of the softs segments on the transition temperature (Ttrans), shape recovery, degradation profile, and drug release kinetics. In vitro study in PBS solution revealed that incorporation of o-PLGA segments to SMPUs is favorable over o-PEG as increased shape memory performance was observed. Moreover, presence of PLGA in PU-PLGA gave more predictable degradation profile in comparison to PU-PLLA/PEG system. Human Cardiac Fibroblasts (HCF) viability tests in vitro confirmed that the amount of Rap released from evaluated PU-PLLA/PEG/Rap and PU-PLGA/Rap drug delivery systems was sufficient to inhibit cells growth on the surface of the tested materials.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Zinn(II)-2-Ethylhexanoat, 92.5-100.0%
Sigma-Aldrich
Glykolid, ≥99%
Sigma-Aldrich
(3S)-cis-3,6-Dimethyl-1,4-dioxan-2,5-dion, 98%