Merck
  • Local release of NECA (5'-(N-ethylcarboxamido)adenosine) from implantable polymeric sheets for enhanced islet revascularization in extrahepatic transplantation site.
Local release of NECA (5'-(N-ethylcarboxamido)adenosine) from implantable polymeric sheets for enhanced islet revascularization in extrahepatic transplantation site.
Journal of controlled release : official journal of the Controlled Release Society (2020-02-23)
Tiep Tien Nguyen, Fakhrossadat Emami, Simmyung Yook, Hanh Thuy Nguyen, Tung Thanh Pham, Shiva Pathak, Shobha Regmi, Jong Oh Kim, Chul Soon Yong, Jae-Ryong Kim, Jee-Heon Jeong
ZUSAMMENFASSUNG
Clinical intraportal pancreatic islet infusion is popular for treating type I diabetes. However, multiple doses of islets and anti-rejection protocols are needed to compensate for early large cell losses post-infusion due to the harsh hepatic environment. Thus, extrahepatic sites are utilized to enable efficient islet engraftment and reduce islet mass. Here, we reported an effective islet revascularization protocol that was based on the co-implantation of islet/fibrin gel construct with poly(lactic-co-glycolic) acid sheet releasing NECA (5'-(N-ethylcarboxamido) adenosine; a potent agonist of adenosine) into mouse epididymal fat pad. Thin, flexible sheets (d = 4 mm) prepared by simple casting exhibited sustained NECA release for up to 21 days, which effectively improved early islet engraftment with a median diabetic reversal time of 18.5 days. Western blotting revealed the facilitative effect of NECA on VEGF expression from islets in vitro and from grafts in vivo. In addition, NECA directly promoted the angiogenic activities of islet-derived endothelial cells by enhancing their proliferation and vessel-like tube formation. As a result, neovasculatures were effectively formed in the engrafted islet vicinity, as evidenced by vasculature imaging and immunofluorescence. Taken together, we suggest NECA-releasing PLGA sheets offer a safe and effective drug delivery system that enhances islet engraftment while reducing islet mass at extrahepatic sites for clinical relevance.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acridinorange -hydrochlorid -Lösung, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin-Fragment 26-33 Amid, ≥97% (HPLC), powder
Sigma-Aldrich
5′-(N-Ethylcarboxamido)adenosine, powder