Merck
Dielectric Property and Space Charge Behavior of Polyimide/Silicon Nitride Nanocomposite Films.
Polymers (2020-02-09)
Minghua Chen, Wenqi Zhou, Jiawei Zhang, Qingguo Chen
ZUSAMMENFASSUNG
Polymeric materials have many applications in multiple industries. In this paper, silicon nitride nanoparticles (Si3N4) were incorporated into a polyimide (PI) matrix to obtain composite films via the in situ polymerization method. The Si3N4 nanoparticles were consistently scattered in the composites, and the thickness of PI/Si3N4 films was around 50 µm. The effects of nanoparticle content on the dielectric constant, loss tangent and breakdown strength were simultaneously studied. A 3 wt.% doped PI/Si3N4 film revealled excellent dielectric properties, a dielectric constant (ε) of 3.62, a dielectric loss tangent (tanδ) of 0.038, and a breakdown strength of 237.42 MV/m. The addition of Si3N4 formed an interface layer inside PI, resulting in a large amount of space charge polarization in the electric field. The space charge of materials from the microscopic point of view was analyzed. The results show that there are trapenergy levels in the composites, which can be used as a composite carrier center and transport channel, effectively improving the performance of a small amount of nanoparticles film.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyromellithsäuredianhydrid, 97%
Sigma-Aldrich
Pyromellitsäure, 96%