Merck
  • Fluorescence Sensing Platforms for Epinephrine Detection Based on Low Temperature Cofired Ceramics.
Fluorescence Sensing Platforms for Epinephrine Detection Based on Low Temperature Cofired Ceramics.
Sensors (Basel, Switzerland) (2020-03-11)
Sylwia Baluta, Karol Malecha, Agnieszka Świst, Joanna Cabaj
ZUSAMMENFASSUNG
A novel fluorescence-sensing pathway for epinephrine (EP) detection was investigated. The ceramic-based miniature biosensor was developed through the immobilization of an enzyme (laccase, tyrosinase) on a polymer-poly-(2,6-di([2,2'-bithiophen]-5-yl)-4-(5-hexylthiophen-2-yl)pyridine), based on low temperature cofired ceramics technology (LTCC). The detection procedure was based on the oxidation of the substrate, i.e., in the presence of the enzyme. An alternative enzyme-free system utilized the formation of a colorful complex between Fe2+ ions and epinephrine molecules. With the optimized conditions, the analytical performance illustrated high sensitivity and selectivity in a broad linear range with a detection limit of 0.14-2.10 nM. Moreover, the strategy was successfully used for an EP injection test with labeled pharmacological samples.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumbisulfat
Sigma-Aldrich
Escherichia coli, Strain K12, lyophilized cells