Merck
  • Farnesoid X receptor (FXR) activation induces the antioxidant protein metallothionein 1 expression in mouse liver.
Farnesoid X receptor (FXR) activation induces the antioxidant protein metallothionein 1 expression in mouse liver.
Experimental cell research (2020-03-08)
Bing Wang, Haibo Zhang, Zhilin Luan, Hu Xu, Yuanyi Wei, Xuejia Zhao, Miaomiao Xing, Xiaoxiao Huo, Jiayang Zhang, Wen Su, Youfei Guan, Xiaoyan Zhang
ZUSAMMENFASSUNG
Farnesoid X receptor (FXR) is a metabolic nuclear receptor, which protects liver from many endogenous and exogenous injuries. Metallothioneins (MTs) belong to a low-molecular-weight protein family involved in metal homeostasis and the regulation of hepatic oxidative stress. In the present study, we aimed to investigate the effect of FXR on hepatic MT1 expression and the underlying mechanism. C57BL/6 mice or primary cultured mouse hepatocytes were treated with the synthetic FXR ligand GW4064 or natural ligand CDCA. RNA-Sequencing (RNA-seq) analysis was performed to identify gene expression profile in the livers of mice treated with GW4064. Real-time PCR and Western blot were applied to determine the expression of MT1 and other FXR target genes in the livers of mice and primary hepatocytes treated with GW4064 and CDCA. Cellular and subcellular locations of MT1 in the livers of mice treated with GW4064 were examined using immunohistochemistry assay. FXR small interfering RNAs (siRNA) was transfected to silence FXR. Luciferase reporter and chromatin immunoprecipitation (ChIP) assays were utilized to confirm the regulation of MT1 gene promoter activity by FXR. RNA-seq analysis revealed that GW4064 treatment significantly induced MT1 expression in mouse liver. Consistently, MT1 expression in the hepatocytes of mouse livers and cultured hepatocytes was upregulated by GW4064 as well as CDCA. In addition, adenovirus-mediated overexpression of FXR markedly increased, while siRNA-mediated FXR silencing significantly suppressed MT1 expression in cultured hepatocytes. Luciferase reporter and ChIP assays further confirmed that the MT1 gene was under the direct control of FXR. Collectively, our findings demonstrate that MT1 is a novel target gene of FXR and may contribute to antioxidative capacity of FXR in liver diseases.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
GW4064, ≥97% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human NR1H4