Merck
  • Structural basis of the interplay between α-synuclein and Tau in regulating pathological amyloid aggregation.
Structural basis of the interplay between α-synuclein and Tau in regulating pathological amyloid aggregation.
The Journal of biological chemistry (2020-04-16)
Jinxia Lu, Shengnan Zhang, Xiaojuan Ma, Chunyu Jia, Zhenying Liu, Chengan Huang, Cong Liu, Dan Li
ZUSAMMENFASSUNG
Amyloid aggregation of pathological proteins is closely associated with a variety of neurodegenerative diseases, and α-synuclein (α-syn) deposition and Tau tangles are considered hallmarks of Parkinson's disease and Alzheimer's disease, respectively. Intriguingly, α-syn and Tau have been found to co-deposit in the brains of individuals with dementia and parkinsonism, suggesting a potential role of cross-talk between these two proteins in neurodegenerative pathologies. Here we show that monomeric α-syn and the two variants of Tau, Tau23 and K19, synergistically promote amyloid fibrillation, leading to their co-aggregation in vitro NMR spectroscopy experiments revealed that α-syn uses its highly negatively charged C terminus to directly interact with Tau23 and K19. Deletion of the C terminus effectively abolished its binding to Tau23 and K19 as well as its synergistic effect on promoting their fibrillation. Moreover, an S129D substitution of α-syn, mimicking C-terminal phosphorylation of Ser129 in α-syn, which is commonly observed in the brains of Parkinson's disease patients with elevated α-syn phosphorylation levels, significantly enhanced the activity of α-syn in facilitating Tau23 and K19 aggregation. These results reveal the molecular basis underlying the direct interaction between α-syn and Tau. We proposed that this interplay might contribute to pathological aggregation of α-syn and Tau in neurodegenerative diseases.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Avanti
18:1 PS (DOPS), Avanti Research - A Croda Brand
Avanti
18:1 PS (DOPS), Avanti Research - A Croda Brand
Avanti
16:0–18:1 PE, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PE, Avanti Research - A Croda Brand