Merck
Interaction of cationic peptides with bacterial membranes.
Methods in molecular biology (Clifton, N.J.) (1997-01-01)
S Fidai, S W Farmer, R E Hancock
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dansyl labeled polymyxin B, Ready Made Solution, for fluorescent microbial imaging, 1.5 mg/mL in H2O