Merck
  • Temperature-responsive self-assembled monolayers of oligo(ethylene glycol): control of biomolecular recognition.
Temperature-responsive self-assembled monolayers of oligo(ethylene glycol): control of biomolecular recognition.
ACS nano (2009-02-12)
Hadi M Zareie, Cyrille Boyer, Volga Bulmus, Ebrahim Nateghi, Thomas P Davis
ZUSAMMENFASSUNG
Self-assembled monolayers (SAMs) of oligo(ethylene glycol) (OEG)-tethered molecules on gold are important for various biorelevant applications ranging from biomaterials to bioanalytical devices, where surface resistance to nonspecific protein adsorption is needed. Incorporation of a stimuli-responsive character to the OEG SAMs enables the creation of nonfouling surfaces with switchable functionality. Here we present an OEG-derived structure that is highly responsive to temperature changes in the vicinity of the physiological temperature, 37 degrees C. The temperature-responsive solution behavior of this new compound was demonstrated by UV-vis and nuclear magnetic resonance spectroscopy. Its chemisorption onto gold(111), and the retention of responsive behavior after chemisorption have been demonstrated by surface plasmon resonance (SPR), X-ray photoelectron spectroscopy (XPS), and atomic force and scanning tunneling microscopy. The OEG-derived SAMs have been shown to reversibly switch the wettability of the surface, as determined by contact angle measurements. More importantly, SPR and AFM studies showed that the OEG SAMs can be utilized to control the affinity binding of streptavidin to the biotin-tethered surface in a temperature-dependent manner while still offering the nonspecific protein-resistance to the surface.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
m-dPEG®36-OH, >90% (HPLC)