Merck
  • Asymmetric catalysis of hetero-ene reactions with tridentate Schiff base chromium(III) complexes.
Asymmetric catalysis of hetero-ene reactions with tridentate Schiff base chromium(III) complexes.
Journal of the American Chemical Society (2002-03-21)
Rebecca T Ruck, Eric N Jacobsen
ZUSAMMENFASSUNG
Tridentate Schiff base chromium(III) complex 1 catalyzes the asymmetric hetero-ene reaction between aryl aldehydes and either 2-methoxypropene or 2-trimethylsilyloxypropene to provide a series of beta-hydroxyenol ether products in high yields and enantioselectivities. X-ray crystallographic analysis of a closely related chromium complex reveals a bridged, dimeric structure bearing aquo-bound six-coordinate Cr(III) centers. A mechanism is proposed wherein water dissociation is effected by means of a chemical desiccant (BaO or silyl enol ether), thereby revealing the site for aldehyde complexation.