- Hypothermia in a patient with Alzheimer's disease receiving a combination therapy with risperidone and pipamperone.
Hypothermia in a patient with Alzheimer's disease receiving a combination therapy with risperidone and pipamperone.
Therapeutic advances in psychopharmacology (2016-02-26)
Daniel Kamp, Myrella Paschali, Tillmann Supprian, Christian Lange-Asschenfeldt
PMID26913178