Merck
  • Soluble protein tyrosine phosphatase receptor type Z (PTPRZ) in cerebrospinal fluid is a potential diagnostic marker for glioma.
Soluble protein tyrosine phosphatase receptor type Z (PTPRZ) in cerebrospinal fluid is a potential diagnostic marker for glioma.
Neuro-oncology advances (2020-07-10)
Yu Yamanoi, Masazumi Fujii, Yuta Murakami, Kenichiro Nagai, Kyoka Hoshi, Yasuhiro Hashimoto, Takashi Honda, Kiyoshi Saito, Shinobu Kitazume
ZUSAMMENFASSUNG
High-grade glioma is the most pervasive and lethal of all brain malignancies. Despite advances in imaging technologies, discriminating between gliomas and other brain diseases such as multiple sclerosis (MS) often requires brain biopsy. Several reports show that protein tyrosine phosphatase receptor Z (PTPRZ) is highly expressed in glioblastoma, and we found that a soluble cleaved form of PTPRZ (sPTPRZ) was present in the cerebrospinal fluid (CSF). The aim of this study was to determine whether the sPTPRZ level in CSF has utility as a diagnostic marker for glioma. Microarray datasets from normal brain tissue and brain tumors were obtained from the Gene Expression Omnibus. PTPRZ protein expression in clinical specimens was evaluated by immunohistochemistry. Semiquantitative western blotting was used to measure sPTPRZ levels in CSF samples from patients with glioma, schwannoma, MS, or nontumor disorders. Expression of PTPRZ mRNA and protein was markedly increased in glioblastoma, astrocytoma, oligodendroglioma, and schwannoma tissues compared with control brain tissue. sPTPRZ was present at significantly elevated levels in the CSF of patients with glioma (grades 1-4), but not in patients with schwannoma or MS, compared with the control samples. Receiver operating characteristic curve analysis showed that sPTPRZ in CSF could discriminate between glioma and MS patients (area under the curve 0.9676; P < .0001). sPTPRZ in CSF is a promising diagnostic biomarker for glioma and could reduce the need for a surgical biopsy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-PTPRZ1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Chondroitin Sulfate Proteoglycan Antibody, Core Protein Epitope, clone Cat-315, ascites fluid, clone Cat-315, Chemicon®