Merck
Crystal structure of ethyl 2-[(4-bromo-phen-yl)amino]-3,4-di-methyl-pent-3-enoate.
Acta crystallographica. Section E, Structure reports online (2014-12-09)
Julio Zukerman-Schpector, I Caracelli, Hélio A Stefani, Amna N Khan, Edward R T Tiekink
ZUSAMMENFASSUNG
In the title compound, C15H20BrNO2, there are two independent mol-ecules (A and B) comprising the asymmetric unit and these adopt very similar conformations. In A, the dihedral angle between the CO2 and MeC=CMe2 groups is 80.7 (3)°, and these make dihedral angles of 3.5 (3) and 84.09 (16)°, respectively, with the bromo-benzene ring. The equivalent dihedral angles for mol-ecule B are 78.4 (3), 2.1 (3) and 78.37 (12)°, respectively. The most prominent inter-actions in the crystal packing are amine-N-H⋯O(carbon-yl) hydrogen bonds between the two independent mol-ecules, resulting in non-centrosymmetric ten-membered {⋯OC2NH}2 synthons. Statistical disorder is noted for each of the terminal methyl groups of the ethyl residues.