Merck
  • A new family of tunable indolylphosphine ligands by one-pot assembly and their applications in Suzuki-Miyaura coupling of aryl chlorides.
A new family of tunable indolylphosphine ligands by one-pot assembly and their applications in Suzuki-Miyaura coupling of aryl chlorides.
The Journal of organic chemistry (2008-09-10)
Chau Ming So, Chung Chiu Yeung, Chak Po Lau, Fuk Yee Kwong
ZUSAMMENFASSUNG
This study describes a new class of indolylphosphine ligands, which can be easily accessed by a simple one-pot assembly from commercially available indoles, acid chlorides, and chlorophosphines. A combination of these three starting materials provides a high diversification of the ligand structure. The application of this ligand array in palladium-catalyzed Suzuki-Miyaura coupling reaction of aryl chlorides with arylboronic acids is described. A catalyst loading down to 0.01 mol % of Pd can be achieved.