Merck
Angiotensin II induces coordinated calcium bursts in aldosterone-producing adrenal rosettes.
Nature communications (2020-04-05)
Nick A Guagliardo, Peter M Klein, Christina A Gancayco, Adam Lu, Sining Leng, Rany R Makarem, Chelsea Cho, Craig G Rusin, David T Breault, Paula Q Barrett, Mark P Beenhakker
ZUSAMMENFASSUNG
Aldosterone-producing zona glomerulosa (zG) cells of the adrenal gland arrange in distinct multi-cellular rosettes that provide a structural framework for adrenal cortex morphogenesis and plasticity. Whether this cyto-architecture also plays functional roles in signaling remains unexplored. To determine if structure informs function, we generated mice with zG-specific expression of GCaMP3 and imaged zG cells within their native rosette structure. Here we demonstrate that within the rosette, angiotensin II evokes periodic Cav3-dependent calcium events that form bursts that are stereotypic in form. Our data reveal a critical role for angiotensin II in regulating burst occurrence, and a multifunctional role for the rosette structure in activity-prolongation and coordination. Combined our data define the calcium burst as the fundamental unit of zG layer activity evoked by angiotensin II and highlight a novel role for the rosette as a facilitator of cell communication.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aldosteron, ≥95% (HPLC)
Sigma-Aldrich
Cyclopiazonsäure aus Penicillium cyclopium, ≥98% (HPLC), powder
Sigma-Aldrich
Minimum Essential Medium Eagle, With Hanks′ salts, L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Anti-N-Cadherin antibody, Mouse monoclonal, clone GC-4, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Pan Cadherin antibody produced in mouse, clone CH-19, ascites fluid
Sigma-Aldrich
N6-Cyclopentyladenosine, solid