Merck
Angiotensin II promotes podocyte injury by activating Arf6-Erk1/2-Nox4 signaling pathway.
PloS one (2020-03-03)
Guanghua Che, Hang Gao, Qibo Hu, Hongchang Xie, Yunfeng Zhang
ZUSAMMENFASSUNG
Angiotensin II (Ang II) is a key contributor to glomerular disease by predominantly resulting in podocyte injury, whereas the underlying molecular mechanisms has not been fully understood. This study aimed to investigate if and how ADP-ribosylation factor 6 (Arf6), a small GTP-binding protein, involves Ang II-induced cellular injury in cultured human podocytes. Cellular injury was evaluated with caspase 3 activity, reactive oxygen species (ROS) level and TUNEL assay. Arf6 activity was measured using an Arf6-GTP Pull-Down Assay. Ang II significantly enhanced Arf6 expressions accompanied by increase of Arf6-GTP. The TUNEL-positive cells as well as activated caspase 3, NADPH oxidase 4 protein (Nox4) and ROS levels were dramatically increased in Ang II-treated podocytes, which was prevented by secinH3, an Arf6 activity inhibitor. Induction of ROS by Ang II was inhibited in podocytes with Nox4 knockdown. Ang II-induced elevation of Nox4 and ROS was prevented by Arf6 knockdown. Phpspho-Erk1/2Thr202/Tyr204 levels were upregulated remarkably following Ang II treatment, and Erk inhibitor LY3214996 significantly downregulated Nox4 expression. In addition, Ang II decreased CD2AP expression. Overexpression of CD2AP prevented Ang II-induced upregulation of Arf6-GTP. Our data demonstrated that Ang II promotes ROS production and podocytes injury through activation of Arf6-Erk1/2-Nox4 signaling. We also provided evidence that Ang II activates Arf6 by degradation of CD2AP.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-15, ascites fluid