Merck
  • Glioma stem-like cells evade interferon suppression through MBD3/NuRD complex-mediated STAT1 downregulation.
Glioma stem-like cells evade interferon suppression through MBD3/NuRD complex-mediated STAT1 downregulation.
The Journal of experimental medicine (2020-03-18)
Xiaoyan Zhan, Saisai Guo, Yuanyuan Li, Haowen Ran, Haohao Huang, Lanjuan Mi, Jin Wu, Xinzheng Wang, Dake Xiao, Lishu Chen, Da Li, Songyang Zhang, Xu Yan, Yu Yu, Tingting Li, Qiuying Han, Kun He, Jiuwei Cui, Tao Li, Tao Zhou, Jeremy N Rich, Shideng Bao, Xuemin Zhang, Ailing Li, Jianghong Man
ZUSAMMENFASSUNG
Type I interferons (IFNs) are known to mediate antineoplastic effects during tumor progression. Type I IFNs can be produced by multiple cell types in the tumor microenvironment; however, the molecular mechanisms by which tumor cells evade the inhibition of immune microenvironment remain unknown. Here we demonstrate that glioma stem-like cells (GSCs) evade type I IFN suppression through downregulation of STAT1 to initiate tumor growth under inhospitable conditions. The downregulation of STAT1 is mediated by MBD3, an epigenetic regulator. MBD3 is preferentially expressed in GSCs and recruits NuRD complex to STAT1 promoter to suppress STAT1 expression by histone deacetylation. Importantly, STAT1 overexpression or MBD3 depletion induces p21 transcription, resensitizes GSCs to IFN suppression, attenuates GSC tumor growth, and prolongs animal survival. Our findings demonstrate that inactivation of STAT1 signaling by MBD3/NuRD provides GSCs with a survival advantage to escape type I IFN suppression, suggesting that targeting MBD3 may represent a promising therapeutic opportunity to compromise GSC tumorigenic potential.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Monoklonaler Anti-α-Tubulin-Antikörper in Maus hergestellte Antikörper, ascites fluid, clone B-5-1-2
Sigma-Aldrich
ChIPAb+-Acetyl-Histon H3 (Lys27) - durch ChIP validiertes Antikörper- und Primer-Set, clone CMA309, from mouse