Merck
  • Efficacy of nitazoxanide, tizoxanide and tizoxanide glucuronide against Cryptosporidium parvum development in sporozoite-infected HCT-8 enterocytic cells.
Efficacy of nitazoxanide, tizoxanide and tizoxanide glucuronide against Cryptosporidium parvum development in sporozoite-infected HCT-8 enterocytic cells.
The Journal of antimicrobial chemotherapy (2000-07-07)
G Gargala, A Delaunay, X Li, P Brasseur, L Favennec, J J Ballet
ZUSAMMENFASSUNG
The effects of nitazoxanide and its metabolites, tizoxanide and tizoxanide glucuronide, on the development of the asexual and sexual stages of Cryptosporidium parvum in differentiated human enterocytic HCT-8 cells were evaluated in a quantitative alkaline phosphatase immunoassay. Nitazoxanide, tizoxanide and tizoxanide glucuronide were inhibitory for up to 46 h when added after sporozoite invasion (MIC50 1.2, 22.6 and 2.2 mg/L, respectively). Tizoxanide had only limited activity, but nitazoxanide and tizoxanide glucuronide strongly inhibited asexual and sexual stages, respectively.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tizoxanide, ≥98% (HPLC)