Merck
  • SYNERGISTIC ON AUXIN AND CYTOKININ 1 positively regulates growth and attenuates soil pathogen resistance.
SYNERGISTIC ON AUXIN AND CYTOKININ 1 positively regulates growth and attenuates soil pathogen resistance.
Nature communications (2020-05-03)
Andrej Hurný, Candela Cuesta, Nicola Cavallari, Krisztina Ötvös, Jerome Duclercq, Ladislav Dokládal, Juan Carlos Montesinos, Marçal Gallemí, Hana Semerádová, Thomas Rauter, Irene Stenzel, Geert Persiau, Freia Benade, Rishikesh Bhalearo, Eva Sýkorová, András Gorzsás, Julien Sechet, Gregory Mouille, Ingo Heilmann, Geert De Jaeger, Jutta Ludwig-Müller, Eva Benková
ZUSAMMENFASSUNG
Plants as non-mobile organisms constantly integrate varying environmental signals to flexibly adapt their growth and development. Local fluctuations in water and nutrient availability, sudden changes in temperature or other abiotic and biotic stresses can trigger changes in the growth of plant organs. Multiple mutually interconnected hormonal signaling cascades act as essential endogenous translators of these exogenous signals in the adaptive responses of plants. Although the molecular backbones of hormone transduction pathways have been identified, the mechanisms underlying their interactions are largely unknown. Here, using genome wide transcriptome profiling we identify an auxin and cytokinin cross-talk component; SYNERGISTIC ON AUXIN AND CYTOKININ 1 (SYAC1), whose expression in roots is strictly dependent on both of these hormonal pathways. We show that SYAC1 is a regulator of secretory pathway, whose enhanced activity interferes with deposition of cell wall components and can fine-tune organ growth and sensitivity to soil pathogens.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Gibberellinsäure, BioReagent, suitable for plant cell culture, ≥90% gibberellin A3 basis (of total gibberellins.)
Sigma-Aldrich
6-Benzylaminopurin, suitable for plant cell culture
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
Proteasehemmer-Cocktail, for plant cell and tissue extracts, DMSO solution
Sigma-Aldrich
Jasmonsäuremethylester, 95%
Sigma-Aldrich
1-Naphthalin-Essigsäure, suitable for plant cell culture, BioReagent, ≥95%, crystalline
Sigma-Aldrich
Monoklonaler Anti-grünfluoreszierendes Protein (GFP)-Antikörper in Maus hergestellte Antikörper, clone GFP-20, ascites fluid
Sigma-Aldrich
24-Epibrassinolid, ≥85%
Supelco
αα-tert-Butyl-β-(4-chlorbenzyl)-1H-1,2,4-triazol-1-ethanol, PESTANAL®, analytical standard
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule), F(ab′)2 fragment–Cy3 antibody produced in sheep, affinity isolated antibody, buffered aqueous solution