Merck
  • KLF4 alleviates cerebral vascular injury by ameliorating vascular endothelial inflammation and regulating tight junction protein expression following ischemic stroke.
KLF4 alleviates cerebral vascular injury by ameliorating vascular endothelial inflammation and regulating tight junction protein expression following ischemic stroke.
Journal of neuroinflammation (2020-04-09)
Xinyu Zhang, Lu Wang, Zhenxiang Han, Jing Dong, Defang Pang, Yuan Fu, Longxuan Li
ZUSAMMENFASSUNG
Although inflammatory cell adhesion molecules (CAMs) and anti-inflammation factor Kruppel-like transcription factor (KLF) 4 have all been reported to be induced after cerebral ischemic stroke (CIS), the close temporal and spatial relationship between expressions of CAMs and KLF4 following CIS and whether and how CAMs and KLF-4 contribute to the development of CIS-induced vascular injury are still unclear. Here, we first examined the correlation between serum levels of CAMs/KLF4 and infarct volume in acute CIS patients. Then, we determined the relationship between CAMs and KLF4 in mice after focal cerebral ischemia. Finally, we investigated the mechanism of KLF4 in protecting against oxygen-glucose deprivation-induced brain endothelial cell injury. Our results demonstrated that patients with moderate to severe CIS had higher serum levels of three CAMs including E-selectin, inter-cellular adhesion molecule 1 (ICAM-1), and vascular cell adhesion molecule 1 (VCAM-1) but lower levels of KLF4 at 48 h after an acute event as compared to patients with minor CIS. The expression levels of three CAMs as well as KLF4 all correlated well with the infarct volume in all the CIS subjects at that time. Although the expressions of three CAMs and KLF4 were all induced in the ischemic hemisphere following focal cerebral ischemia, the peak timing and distribution patterns of their expression were different: the induction of KLF4 lagged behind that of the CAMs in the ischemic penumbra; furthermore, the dual immunofluorescent studies displayed that high expression of KLF4 was always associated with relatively less cerebral vascular endothelial inflammation response in the ischemic hemisphere and vice versa. Mechanistic analyses revealed that KLF4 alleviated CIS-induced cerebral vascular injury by regulating endothelial expressions of CAMs, nuclear factor-kB, and tight junction proteins. These data indicate that KLF4 confers vascular protection against cerebral ischemic injury, suggesting that circulating CAMs and KLF4 might be used as potential biomarkers for predicting the prognosis of acute ischemic stroke and also providing a new proof of concept and potential targets for future prevention and treatment of CIS.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-ZO-1-Antikörper, Klon R40.76, clone R40.76, from rat