Merck
Down-regulation of MTHFD2 inhibits NSCLC progression by suppressing cycle-related genes.
Journal of cellular and molecular medicine (2019-11-30)
Chang Yu, Lehe Yang, Mengsi Cai, Feng Zhou, Sisi Xiao, Yaozhe Li, Tingting Wan, Dezhi Cheng, Liangxing Wang, Chengguang Zhao, Xiaoying Huang
ZUSAMMENFASSUNG
Methylenetetrahydrofolate dehydrogenase 2 (MTHFD2) is a bifunctional enzyme located in the mitochondria. It has been reported to be overexpressed in several malignancies. However, the relationship between the expression of MTHFD2 and non-small cell lung cancer (NSCLC) remains largely unknown. In this study, we found that MTHFD2 was significantly overexpressed in NSCLC tissues and cell lines. Knockdown of MTHFD2 resulted in reduced cell growth and tumorigenicity in vitro and in vivo. Besides, the mRNA and protein expression level of cell cycle genes, such as CCNA2, MCM7 and SKP2, was decreased in MTHFD2 knockdown H1299 cells. Our results indicate that the inhibitory effect of MTHFD2 knockdown on NSCLC may be mediated via suppressing cell cycle-related genes. These findings delineate the role of MTHFD2 in the development of NSCLC and may have potential applications in the treatment of NSCLC.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-MTHFD2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution