Merck
  • Extraction of caffeine, chlorogenic acids and lipids from green coffee beans using supercritical carbon dioxide and co-solvents
Extraction of caffeine, chlorogenic acids and lipids from green coffee beans using supercritical carbon dioxide and co-solvents
Brazilian Journal of Chemical Engineering (2008)
Azevedo dABA, et al.