Merck
  • p-Type semiconducting nickel oxide as an efficiency-enhancing anode interfacial layer in polymer bulk-heterojunction solar cells
p-Type semiconducting nickel oxide as an efficiency-enhancing anode interfacial layer in polymer bulk-heterojunction solar cells
Proceedings of the National Academy of Sciences of the USA (2008)
Irwin, M. D.; et al.