Merck
  • Spin-label-induced selective carbon-13 nuclear relaxation: structures of the. gamma.-cyclodextrin-tempone and. alpha.-cyclodextrin-di-tert-butyl nitroxide inclusion complexes in solution
Spin-label-induced selective carbon-13 nuclear relaxation: structures of the. gamma.-cyclodextrin-tempone and. alpha.-cyclodextrin-di-tert-butyl nitroxide inclusion complexes in solution
Macromolecules (1989)
Eastman MP, et al.