Merck
  • Cationic ring-opening polymerization of carbazolyl-containing epoxy monomers by triphenyl carbenium salts as thermal-and photolatent initiators
Cationic ring-opening polymerization of carbazolyl-containing epoxy monomers by triphenyl carbenium salts as thermal-and photolatent initiators
Monatshefte fur Chemie / Chemical Monthly (2009)
Lazauskaite R, et al.