Merck
  • DNA fromMycobacterium tuberculosisIdentified in Mediaeval Human Skeletal Remains Using Polymerase Chain Reaction
DNA fromMycobacterium tuberculosisIdentified in Mediaeval Human Skeletal Remains Using Polymerase Chain Reaction
Journal of Archaeological Science (1996)
Taylor M G, et al.