Merck
  • Identification of the 4-Position of 3-Alkynyl and 3-Heteroaromatic Substituted Pyridine Methanamines as a Key Modification Site Eliciting Increased Potency and Enhanced Selectivity for Cytochrome P-450 2A6 Inhibition.
Identification of the 4-Position of 3-Alkynyl and 3-Heteroaromatic Substituted Pyridine Methanamines as a Key Modification Site Eliciting Increased Potency and Enhanced Selectivity for Cytochrome P-450 2A6 Inhibition.
Journal of medicinal chemistry (2018-07-12)
Travis T Denton, Pramod Srivastava, Zuping Xia, Gang Chen, Christy J W Watson, Alec Wynd, Philip Lazarus
ZUSAMMENFASSUNG
Cigarette smoking causes nearly one in every five deaths in the United States. The development of a specific inhibitor of cytochrome P450 2A6 (CYP2A6), the major nicotine-metabolizing enzyme in humans, which could be prescribed for the cessation of cigarette smoking, has been undertaken. To further refine the structure activity relationship of CYP2A6, previously synthesized 3-alkynyl and 3-heteroaromatic substituted pyridine methanamines were used as lead compounds. Isosteric pyridine replacement and appendage of all available positions around the pyridine ring with a methyl group was performed to identify a modification that would increase CYP2A6 inhibition potency, which led to 4g (IC50 = 0.055 mM) as a primary analogue. Potent compounds were evaluated for CYP selectivity, human liver microsomal half-life, and compliance with the rules of five. Top compounds (i.e., 6i, IC50 = 0.017 mM, >120 min half-life) are poised for further development as treatments for smoking and tobacco use cessation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DLCI-1, ≥98% (HPLC)