Merck
  • Molecular orbital study of 8-azaxanthine derivatives and crystal structure of 1,3-dimethyl-8-azaxanthine monohydrate
Molecular orbital study of 8-azaxanthine derivatives and crystal structure of 1,3-dimethyl-8-azaxanthine monohydrate
Journal of Molecular Structure (1995)
Purificacion Sanchez, M., et al.