Merck
[The preparation and identification of rabbit's antibody of chymopapain].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2003-04-11)
Sufang Wang, Taicheng Yang, Linlan Jiang, Shujin Zhao
ZUSAMMENFASSUNG
Chymopapain, one of the four cysteine proteinases of papaya latex, has milk clotting and proteolytic activity. It is mainly used to treat prolapsed intervertebral discs. This paper introduced the preparation of polyclonal antibody of partially purified chymopapain and the identification of the antibody specificity. The polyclonal antibody provides a basis for the further researches and applications of chymopapain.