Merck
  • Design, synthesis and structure activity relationships of indazole and indole derivatives as potent glucagon receptor antagonists
Design, synthesis and structure activity relationships of indazole and indole derivatives as potent glucagon receptor antagonists
Bioorganic & Medicinal Chemistry Letters (2019)
Song F, et al.