Merck
  • Validation of a FT-IR method for the determination of oils and grease in water using tetrachloroethylene as the extraction solvent
Validation of a FT-IR method for the determination of oils and grease in water using tetrachloroethylene as the extraction solvent
Desalination (2007)
Farmaki E, et al.