Merck
Bcl-x(L) forms an ion channel in synthetic lipid membranes.
Nature (1997-01-23)
A J Minn, P Vélez, S L Schendel, H Liang, S W Muchmore, S W Fesik, M Fill, C B Thompson
ZUSAMMENFASSUNG
Bcl-2-related proteins are critical regulators of cell survival that are localized to the outer mitochondrial, outer nuclear and endoplasmic reticulum membranes. Despite their physiological importance, the biochemical function of Bcl-2-related proteins has remained elusive. The three-dimensional structure of Bcl-xL, an inhibitor of apoptosis, was recently shown to be similar to the structures of the pore-forming domains of bacterial toxins. A key feature of these pore-forming domains is the ability to form ion channels in biological membranes. Here we demonstrate that Bcl-xL shares this functional feature. Like the bacterial toxins, Bcl-xL can insert into either synthetic lipid vesicles or planar lipid bilayers and form an ion-conducting channel. This channel is pH-sensitive and becomes cation-selective at physiological pH. The ion-conducting channel(s) formed by Bcl-xL display multiple conductance states that have identical ion selectivity. Together, these data suggest that Bcl-xL may maintain cell survival by regulating the permeability of the intracellular membranes to which it is distributed.