Merck
Unique murine tumor-associated antigens identified by tumor infiltrating lymphocytes.
Journal of immunology (Baltimore, Md. : 1950) (1990-02-15)
R J Barth, S N Bock, J J Mulé, S A Rosenberg
ZUSAMMENFASSUNG
Tumor infiltrating lymphocytes (TIL) were cultured from multiple methylcholanthrene-induced sarcomas under four different conditions: either low-dose (10 U/ml) or high-dose (1000 U/ml) rIL-2 was used with Ag stimulation by irradiated autologous tumor and splenocytes starting either at day 1 or day 10 of culture. TIL grown from four antigenically distinct sarcomas in low-dose rIL-2 were specifically lytic in vitro to their tumor of origin in 13 of 15 (87%) lytic cultures whereas only 8 of 28 (29%) lytic TIL cultures grown in high-dose rIL-2 showed specificity. TIL cultured in low-dose rIL-2 with Ag stimulation on day 1 of culture proliferated at a rate equal to TIL grown in high-dose rIL-2 and maintained their specificity for over 3 mo in culture. Cytolysis by specific TIL was MHC-restricted. TIL with in vitro specificity were therapeutically effective in eliminating established micrometastases in murine models. These investigations demonstrate that CTL derived from tumor-bearing mice can be used to define unique tumor-associated Ag on at least four different sarcomas and may be valuable in studies of the biologic nature of these Ag and in the adoptive immunotherapy of tumors.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MCA205-Fibrosarkom-Zelllinie, murin, MCA205 mouse fibrosarcoma cell line is an excellent model for studying immune response to tumor cells and the development of targeted cancer immunotherapies.