Merck
  • Determination of ultra trace arsenic species in water samples by hydride generation atomic absorption spectrometry after cloud point extraction.
Determination of ultra trace arsenic species in water samples by hydride generation atomic absorption spectrometry after cloud point extraction.
Analytica chimica acta (2011-09-06)
Halil İbrahim Ulusoy, Mehmet Akçay, Songül Ulusoy, Ramazan Gürkan
ZUSAMMENFASSUNG
Cloud point extraction (CPE) methodology has successfully been employed for the preconcentration of ultra-trace arsenic species in aqueous samples prior to hydride generation atomic absorption spectrometry (HGAAS). As(III) has formed an ion-pairing complex with Pyronine B in presence of sodium dodecyl sulfate (SDS) at pH 10.0 and extracted into the non-ionic surfactant, polyethylene glycol tert-octylphenyl ether (Triton X-114). After phase separation, the surfactant-rich phase was diluted with 2 mL of 1M HCl and 0.5 mL of 3.0% (w/v) Antifoam A. Under the optimized conditions, a preconcentration factor of 60 and a detection limit of 0.008 μg L(-1) with a correlation coefficient of 0.9918 was obtained with a calibration curve in the range of 0.03-4.00 μg L(-1). The proposed preconcentration procedure was successfully applied to the determination of As(III) ions in certified standard water samples (TMDA-53.3 and NIST 1643e, a low level fortified standard for trace elements) and some real samples including natural drinking water and tap water samples.