Merck
  • Tetrabromobisphenol A inhibits carboxylesterase activity of marine organisms from different trophic levels.
Tetrabromobisphenol A inhibits carboxylesterase activity of marine organisms from different trophic levels.
Chemosphere (2019-08-24)
David Nos, Joan Navarro, Enric Saiz, Juan C Sanchez-Hernandez, Montserrat Solé
ZUSAMMENFASSUNG
Tetrabromobisphenol A (TBBPA), a brominated flame retardant used in synthetic polymers and electronics, is present in the aquatic environment and recent evidence suggests it can be potentially biomagnified in the marine ecosystem. However, the toxicity of TBBPA in the marine biota has not been investigated in detail. In this study we aimed to understand the role of carboxylesterases (CEs) in xenobiotic metabolism under the exposure of marine organisms to a chemical of environmental concern, TBBPA. Specifically, we tested for in vitro inhibition of CE activity in a range of marine organisms covering different ecological niches, from species from low (mussels and copepods), medium (sardines and anchovies) and high trophic levels (tuna). The results revealed that the highest inhibition of CE activity to 100 μM TBBPA was recorded in mussels (66.5% inhibition) and tunids (36.3-76.4%), whereas copepods and small pelagic fish showed comparatively lower effects (respectively, 30% and 36.5-55.6%). Our results suggest that CE-mediated detoxification and physiological processes could be compromised in TBBPA-exposed organisms and could ultimately affect humans as many of them are market species.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Carboxylesterase 2 human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Carboxylesterase 1 isoform c human, recombinant, expressed in baculovirus infected BTI insect cells