- Aggregation-Induced Emission: Together We Shine, United We Soar!
Aggregation-Induced Emission: Together We Shine, United We Soar!
Chemical reviews (2015-10-23)
Ju Mei, Nelson L C Leung, Ryan T K Kwok, Jacky W Y Lam, Ben Zhong Tang
PMID26492387
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
LuminiCell Tracker 670- Cell Labeling Kit, Biocompatible organic fluorescent nanoparticles (AIEDots) that can be used to label cells for long term cell tracking and cell tracing in vitro and in vivo experiments.
Sigma-Aldrich
LuminiCell Tracker 540- Cell Labeling Kit, Biocompatible organic fluorescent nanoparticles (AIEDots) that can be used to label cells for long term cell tracking and cell tracing in vitro and in vivo experiments.
Sigma-Aldrich
LuminiCell Tracker 540- Vascular Labeling Kit, Biocompatible organic fluorescent nanoparticles (AIEDots) that can be used to fluorescently tag vasculature in living tissues and animals for studies of inflammation and vascular leakage.
Sigma-Aldrich
LuminiCell Tracker 670- Vascular Labeling Kit, Biocompatible organic fluorescent nanoparticles (AIEDots) that can be used to fluorescently tag vasculature in living tissues and animals for studies of inflammation and vascular leakage.