Merck
  • ALS2CL, the novel protein highly homologous to the carboxy-terminal half of ALS2, binds to Rab5 and modulates endosome dynamics.
ALS2CL, the novel protein highly homologous to the carboxy-terminal half of ALS2, binds to Rab5 and modulates endosome dynamics.
FEBS letters (2004-09-25)
Shinji Hadano, Asako Otomo, Kyoko Suzuki-Utsunomiya, Ryota Kunita, Yoshiko Yanagisawa, Junko Showguchi-Miyata, Hikaru Mizumura, Joh-E Ikeda
ZUSAMMENFASSUNG
ALS2, the causative gene product for juvenile recessive amyotrophic lateral sclerosis (ALS2), is a guanine-nucleotide exchange factor for the small GTPase Rab5. Here, we report a novel ALS2 homologous gene, ALS2 C-terminal like (ALS2CL), which encodes a 108-kD ALS2CL protein. ALS2CL exhibited a specific but a relatively weak Rab5-GEF activity with accompanying rather strong Rab5-binding properties. In HeLa cells, co-expression of ALS2CL and Rab5A resulted in a unique tubulation phenotype of endosome compartments with significant colocalization of ALS2CL and Rab5A. These results suggest that ALS2CL is a novel factor modulating the Rab5-mediated endosome dynamics in the cells.