Merck
  • Polymer brushes on carbon nanotubes by thiol-lactam initiated radical polymerization of 2-hydroxyethyl methacrylate.
Polymer brushes on carbon nanotubes by thiol-lactam initiated radical polymerization of 2-hydroxyethyl methacrylate.
Journal of nanoscience and nanotechnology (2012-04-25)
Md Harun-Or Rashid, Won-Ki Lee, Seong-Soo Hong, Jong Myung Park, Hyun Gyu Kim, Kwon Taek Lim
ZUSAMMENFASSUNG
Water-soluble polymer brushes with multi-walled carbon nanotubes (MWNTs) as backbones were synthesized by grafting 2-hydroxyethyl methacrylate (HEMA) from surface functionalized MWNTs via in situ surface thiol-lactam initiated radical polymerization. MWNTs were functionalized with 2-mercaptoethanol and used as initiators in the polymerization of HEMA in the presence of butyrolactam. FT-IR, XPS, 1H NMR, GPC and TGA were used to determine chemical structure and the grafted polymer quantities of the resulting product. The covalent bonding of PHEMA to the MWNTs dramatically improved the water dispersibility of MWNTs. The average thicknesses of the polymer brushes in the functionalized MWNTs were detected with electron microscopy (SEM and TEM) and images indicated that the nanotubes were coated with polymer layer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Hydroxyethylmethacrylat