Merck
Influence of sickle cell disease on susceptibility to HIV infection.
PloS one (2020-04-09)
Shannon Kelly, Evan S Jacobs, Mars Stone, Sheila M Keating, Tzong-Hae Lee, Daniel Chafets, John Heitman, Melanie Dimapasoc, Eva Operskalski, Ward Hagar, Elliott Vichinsky, Michael P Busch, Philip J Norris, Brian Custer
ZUSAMMENFASSUNG
People with sickle cell disease (SCD) are reported to have low rates of HIV infection, slower progression to AIDS and lower HIV-associated mortality compared to the general population. Mechanisms of potential resistance to HIV in SCD are incompletely understood. We retrospectively reviewed the Transfusion Safety Study to compare HIV status between people with SCD and other congenital anemias who were routinely exposed to blood products during the high-risk period before HIV screening implementation. Non-SCD congenital anemia diagnosis was associated with a higher risk of HIV acquisition compared to SCD (OR 13.1 95%CI 1.6-108.9). In addition, we prospectively enrolled 30 SCD cases and 30 non-SCD controls to investigate potential mechanisms of resistance to HIV in SCD. CCR5 and CCR7 expression was lower and CD4 expression was higher on CD4+ T cells from SCD cases compared to controls. Surface expression of CD4+ T cell CXCR4, CD38 and HLA-DR did not differ between the groups. SCD CD4+ T cells were not less susceptible to HIV infection than controls. Levels of multiple cytokines were elevated in the SCD plasma, but SCD plasma compared to control plasma did not inhibit HIV infection of target cells. In conclusion, our epidemiological data support people with SCD being resistant to HIV infection. Potential mechanisms include lower CD4+ T cell expression of CCR5 and CCR7, balanced by increased CD4 expression and cytokine levels, which did not result in in vitro resistance to HIV infection. Further study is needed to define the risk and pathophysiology of HIV in persons with SCD.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acridinorange -hydrochlorid -Lösung, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
N-Acetyl-D-penicillamin, ≥99.0% (T)