Merck
  • Revision of Telothyria van der Wulp (Diptera: Tachinidae) and twenty-five new species from Area de Conservación Guanacaste in northwestern Costa Rica with a key to Mesoamerican species.
Revision of Telothyria van der Wulp (Diptera: Tachinidae) and twenty-five new species from Area de Conservación Guanacaste in northwestern Costa Rica with a key to Mesoamerican species.
Biodiversity data journal (2020-05-12)
A J Fleming, D Monty Wood, M Alex Smith, Tanya Dapkey, Winnie Hallwachs, Daniel Janzen
ZUSAMMENFASSUNG
We describe 25 new species in the genus Telothyria van der Wulp, 1890 from Area de Conservación Guanacaste (ACG) in northwestern Costa Rica. All species herein described were reared from an ongoing inventory of wild-caught caterpillars spanning two families (Lepidoptera: Crambidae, and Tortricidae). Our study provides a concise description of each new species using morphology, life history, molecular data, and photographic documentation; a redescription of the genus, and its type species as well as a revised key to species of Telothyria occurring in the Mesoamerican region. We also suggest seven new synonymies resulting in 11 new combinations. The following 25 new species of Telothyria are described: T. aidanisp. n., T. alexanderisp. n., T. auranticrussp. n., T. auriolussp. n., T. bicuspidatasp. n., T. carolinacanoaesp. n., T. clavatasp. n., T. cristatasp. n., T. diniamartinezaesp. n., T. duniagarciaesp. n., T. duvalierbricenoisp. n., T. eldaarayaesp. n., T. erythropygasp. n., T. fimbriatasp. n., T. fulgidasp. n., T. gloriashihezaraesp. n., T. griseasp. n., T. harryramirezisp. n., T. incisasp. n., T. manuelpereiraisp. n., T. obscurasp. n., T. omissasp. n., T. osvaldoespinozaisp. n., T. peltatasp. n., and T. ricardocaleroisp. n.The following are proposed by Fleming & Wood as new generic synonyms of Telothyria: Comatacta Coquillett Syn. n., Floradalia Thompson Syn. n., Ptilomyia Curran Syn. n., Ptilomyiopsis Townsend Syn. n., Ptilomyoides Curran Syn. n., EuptilomyiaSyn. n., Eutelothyria Townsend Syn. n.The following new combinations are proposed as a result of the new synonymies: Telothyria bequaerti (Curran, 1925) Comb. n., Telothyria cruenta (Giglio-Tos, 1893) Comb. n., Telothyria frontalis (Townsend, 1939) Comb. n., Telothyria insularis (Curran, 1927) Comb. n., Telothyria itaquaquecetubae (Townsend, 1931) Comb. n., Telothyria major (Thompson, 1963) Comb. n., Telothyria micropalpus (Curran, 1925) Comb. n., Telothyria minor (Thompson, 1963) Comb. n., Telothyria nautlana (Townsend, 1908) Comb. n., Telothyria plumata (Curran, 1925) Comb. n., Telothyria trinitatis (Thompson, 1963) Comb. n., Telothyria variegata (Fabricius, 1805) Comb. n.Musca tricincta Fabricius is synonymized under Telothyria variegata Fabricius, Syn. n.Telothyria schineri Fleming & Wood nom. n. is proposed as a replacement name for Miltogramma brevipennis Schiner.Additionally we provide redescriptions of two previously named species: the type species Telothyria cupreiventris (van der Wulp) due to its being the type species, and Telothyria relicta (van der Wulp) due to its having been reared as an outcome of the inventory.