Merck
  • A DEAD-box RNA helicase TCD33 that confers chloroplast development in rice at seedling stage under cold stress.
A DEAD-box RNA helicase TCD33 that confers chloroplast development in rice at seedling stage under cold stress.
Journal of plant physiology (2020-03-28)
Wang Xiaomei, Kong Rongrong, Zhang Ting, Gao Yuanyuan, Xu Jianlong, Piao Zhongze, Lee Gangseob, Lin Dongzhi, Dong Yanjun
ZUSAMMENFASSUNG
Cold stress is one of the most common unfavorable environmental factors affecting the growth, development, and survival of plants. The DEAD-box RNA helicases play important roles in all types of processes of RNA metabolism. However, the function of DEAD-box RNA helicase under cold stress is poorly explored in plants, especially in rice. This study reported the identification of a novel rice thermo-sensitive chlorophyll-deficient mutant, tcd33, which displayed an albino phenotype before the four-leaf stage, then withered and eventually died at 20 °C, while wild-type plants exhibited normal green coloration at 32 °C. The tcd33 seedlings also exhibited less chlorophyll contents and severe defects of chloroplast structure under 20 °C condition. Map-based cloning and complementation experiments suggested that TCD33 encodes a chloroplast-located DEAD-box RNA helicase protein. The transcript expression level of TCD33 indicated that the genes related to chlorophyll (Chl) biosynthesis, photosynthesis, and chloroplast development in tcd33 mutants were down-regulated at 20 °C, while the down-regulated genes were nearly recovered to or slightly higher than the WT level at 32 °C. Together, our results suggest that the cold-inducible TCD33 is essential for early chloroplast development and is important for cold-responsive gene regulation and cold tolerance in rice.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Chlorophyll b, analytical standard