Merck
Circumvention of common labelling artefacts using secondary nanobodies.
Nanoscale (2020-05-02)
Shama Sograte-Idrissi, Thomas Schlichthaerle, Carlos J Duque-Afonso, Mihai Alevra, Sebastian Strauss, Tobias Moser, Ralf Jungmann, Silvio O Rizzoli, Felipe Opazo
ZUSAMMENFASSUNG
A standard procedure to study cellular elements is via immunostaining followed by optical imaging. This methodology typically requires target-specific primary antibodies (1.Abs), which are revealed by secondary antibodies (2.Abs). Unfortunately, the antibody bivalency, polyclonality, and large size can result in a series of artifacts. Alternatively, small, monovalent probes, such as single-domain antibodies (nanobodies) have been suggested to minimize these limitations. The discovery and validation of nanobodies against specific targets are challenging, thus only a minimal amount of them are currently available. Here, we used STED, DNA-PAINT, and light-sheet microscopy, to demonstrate that secondary nanobodies (1) increase localization accuracy compared to 2.Abs; (2) allow direct pre-mixing with 1.Abs before staining, reducing experimental time, and enabling the use of multiple 1.Abs from the same species; (3) penetrate thick tissues more efficiently; and (4) avoid probe-induced clustering of target molecules observed with conventional 2.Abs in living or poorly fixed samples. Altogether, we show how secondary nanobodies are a valuable alternative to 2.Abs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Maleimid, 99%
Sigma-Aldrich
Dibenzocyclooctyn-sulfo-N-hydroxysuccinimidyl-ester, for Copper-free Click Chemistry
Sigma-Aldrich
DL-Cystein, technical grade