Merck
  • Recombinant spider silk protein eADF4(C16)-RGD coatings are suitable for cardiac tissue engineering.
Recombinant spider silk protein eADF4(C16)-RGD coatings are suitable for cardiac tissue engineering.
Scientific reports (2020-05-31)
Johannes P M Kramer, Tamara B Aigner, Jana Petzold, Kaveh Roshanbinfar, Thomas Scheibel, Felix B Engel
ZUSAMMENFASSUNG
Cardiac tissue engineering is a promising approach to treat cardiovascular diseases, which are a major socio-economic burden worldwide. An optimal material for cardiac tissue engineering, allowing cardiomyocyte attachment and exhibiting proper immunocompatibility, biocompatibility and mechanical characteristics, has not yet emerged. An additional challenge is to develop a fabrication method that enables the generation of proper hierarchical structures and constructs with a high density of cardiomyocytes for optimal contractility. Thus, there is a focus on identifying suitable materials for cardiac tissue engineering. Here, we investigated the interaction of neonatal rat heart cells with engineered spider silk protein (eADF4(C16)) tagged with the tripeptide arginyl-glycyl-aspartic acid cell adhesion motif RGD, which can be used as coating, but can also be 3D printed. Cardiomyocytes, fibroblasts, and endothelial cells attached well to eADF4(C16)-RGD coatings, which did not induce hypertrophy in cardiomyocytes, but allowed response to hypertrophic as well as proliferative stimuli. Furthermore, Kymograph and MUSCLEMOTION analyses showed proper cardiomyocyte beating characteristics on spider silk coatings, and cardiomyocytes formed compact cell aggregates, exhibiting markedly higher speed of contraction than cardiomyocyte mono-layers on fibronectin. The results suggest that eADF4(C16)-RGD is a promising material for cardiac tissue engineering.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fibronektin-Rinderplasma, solution, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
(+)-Natrium-L-ascorbat, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Pancreatin aus Schweinepankreas, powder, suitable for cell culture, 4 × USP specifications
Sigma-Aldrich
Albumin -Lösung, 35% in 0.85% sodium chloride, aseptically filled
Sigma-Aldrich
(R)-(−)-Phenylephrin -hydrochlorid, powder