Merck
  • Erythropoietin is a major regulator of thrombopoiesis in thrombopoietin-dependent and -independent contexts.
Erythropoietin is a major regulator of thrombopoiesis in thrombopoietin-dependent and -independent contexts.
Experimental hematology (2020-07-30)
Salima Hacein-Bey-Abina, Machadiya Estienne, Stéphanie Bessoles, Hamid Echchakir, Magali Pederzoli-Ribeil, Andrada Chiron, Lydia Aldaz-Carroll, Valentin Leducq, Yanyan Zhang, Michèle Souyri, Fawzia Louache, Amine M Abina
ZUSAMMENFASSUNG
Thrombopoietin (TPO), through activation of its cognate receptor Mpl, is the major regulator of platelet production. However, residual platelets observed in TPO- and Mpl-loss-of-function (LOF) mice suggest the existence of an additional factor to TPO in platelet production. As erythropoietin (EPO) exhibited both in vitro megakaryocytic potential, in association with other early-acting cytokines, and in vivo platelet activation activity, we sought to investigate its role in this setting. Here, we used multiple LOF models to decipher the reciprocal role of EPO and TPO in the regulation of platelet production in TPO-LOF and Mpl-LOF mice and of platelet size heterogeneity in wild-type mice. We first identified EPO as the major thrombopoietic factor in the absence of the TPO-Mpl pathway. Based on the study of several mouse models we found that the EPO-EPO receptor pathway acts on late-stage megakaryopoiesis and is responsible for large-sized platelet production, while the TPO-Mpl pathway promotes small-sized platelet production. On the basis of our data, EPO might be used for thrombocytopenia supportive therapy in congenital amegakaryocytopoiesis. Furthermore, as a distribution skewed toward large platelets is an independent risk factor and a poor prognosis indicator in atherothrombosis, the characterization of EPO's role in the production of large-sized platelets, if confirmed in humans, may open new perspectives in the understanding of the role of EPO-induced platelets in atherothrombosis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Histopaque®-1077, sterile-filtered, density: 1.077