Merck
Idronoxil as an Anticancer Agent: Activity and Mechanisms.
Current cancer drug targets (2020-01-04)
Kate Porter, Walter Douglas Fairlie, Olivier Laczka, Frederic Delebecque, John Wilkinson
ZUSAMMENFASSUNG
Idronoxil has been the subject of more than 50 peer-reviewed publications over the last two decades. This isoflavone is an intriguing regulator of multiple signal transduction pathways, capable of causing a range of biological effects, including cell cycle arrest, apoptosis, an ability to stimulate the immune system, and inhibition of angiogenesis. These multifaceted actions suggest that idronoxil has the potential to synergize with, or complement, a wide range of cancer therapies. Whilst clinically tested in the past, idronoxil's journey was discontinued as a result of its low bioavailability in humans when administered either intravenously or orally, though strategies to overcome this issue are currently being explored. Here, we summarize the current literature regarding the key cellular targets of idronoxil and the mechanisms by which idronoxil exerts its anticancer effects, laying a new foundation toward giving this unique molecule a second chance of contributing to the future of cancer treatment.